missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ Polydisc In-line Filter
For serum samples
$248.00 - $827.00
Catalog Number | Mfr. No. | Material | Pore Size | Flow Rate | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Pore Size | Flow Rate | Quantity | Price | Quantity | ||
09744105
|
Cytiva
67245000 |
PES | 1.0 μm | 500.0mL/min./cm sq. | 10/Pk. |
Pack of 10 Each for $248.00
|
|
||
09-744-84
|
Cytiva
67245002 |
PES | 0.2 μm | 500.0mL/min./cm sq. | 10/Pk. |
Pack of 10 Each for $254.00
|
|
||
09927227
|
Cytiva
2227 |
Polypropylene | 5 μm | 1500mL/min | 50 |
Pack of 50 Each for $827.00
|
|
||
Description
- Glass microfiber and polyethersulfone membranes
- Contains stack of filter media
- Radiation sterilized