missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Polycap™ TC High-Capacity Capsules

Developed for safe, efficient filtration of critical biological and tissue culture solutions. Whatman™ Polycap™ TC High-Capacity Capsules include PES membranes for excellent flow rate and protein absorption characteristics.

$148.84 - $259.80

Specifications

Material (Membrane) Polyethersulfone (PES)
Pore Size 0.8; 0.2μm
Housing Material Polypropylene
Outlet Style Stepped barb for 6 to 10mm tubing
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Filtration Area (Metric) Inlet Style Price Quantity  
Catalog Number Mfr. No. Filtration Area (Metric) Inlet Style Price Quantity  
05-713-750 Whatman™
67153682
440cm2 Stepped barb for 6 to 10mm tubing Pack for $148.84
05-713-751 Whatman™
67157582
930cm2 Stepped barb for 6 to 10mm tubing Pack for $228.09
05-713-752 Whatman™
67189582
1900cm2 Stepped barb for 10 to 12mm tubing Pack for $259.80
Specifications

Description

Contain two layers of polyethersulfone membrane — inherently hydrophilic, low in extractables, excellent flowrates. Low protein-binding characteristics make capsules ideal for use with biologically sensitive solutions.

  • Pore size 0.8/0.2μm
  • Filtration area: 440, 930, or 1900cm2
  • Vent evacuates any air for fast filtration
  • Outlet is protected to prevent touch contamination during setup
  • Standard hose barb connections
  • Durable polypropylene filter housing can withstand maximum operating pressure of 4.1 bar (60psi)
  • Presterilized ISO-driven cleanroom manufacturing ensures product consistency
  • Stamped lot numbers ensure traceability of all critical components

Suitable for pharmaceutical aqueous solutions, tissue culture media, reagent preparation, virus suspensions, particle counting solutions, cleaning and rinsing solutions.

Compliance

Pass HIMA Challenge Test for Sterilizing Grade Filters to retain >107CFU/cm2 Brevundimonas diminuta per ASTM™ 838-83 and FDA guidelines; microbial retention correlated to 100% integrity testing during manufacturing.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

Polyethersulfone (PES)
0.8; 0.2μm
Polypropylene
Stepped barb for 6 to 10mm tubing