Please login to your online account to display your discounted pricing
Polycap GW 75 High-Capacity Capsules
Filter GW (Ground Water) samples before dissolved metal analysis
Manufacturer: ge healthcare - whatman  WHT67246004
 View more versions of this product
Catalog No. 05-714-037
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
Quantity 100/pk.
Material (Membrane) Polyethersulfone
Certifications/Compliance Certified for low metal extractables; Suitable for EPA Method 3005 for groundwater analysis
Filtration Area (Metric) 600cm2
Sterile No
Housing Material Polypropylene
Pore Size 0.45μm