missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Whatman™ Polycap Disposable Capsules: 75TC

Polycap TC filters for sterile filtration of biological and tissue culture aqueous solutions

$178.66 - $182.94

Specifications

Material (Membrane) Polyethersulfone
Filtration Area 930 cm2
For Use With (Application) Sterile filtration of biological and tissue culture solutions. Principally for aqueous solutions.
Sterility Sterile
View More Specs

Products 5
Catalog Number Mfr. No. Pore Size Inlet Style Outlet Style Price Quantity  
Catalog Number Mfr. No. Pore Size Inlet Style Outlet Style Price Quantity  
09-747-6
View Documents
Cytiva
67147502
0.2/0.2 μm 1/4-3/8 in. with SB inlet and outlet barb 1/4-3/8 in. with SB inlet and outlet barb
Pack for $178.66
 
09-747-15
View Documents
Cytiva
67177504
0.6/0.45 μm 3/8-1/2 in. with 1/2 SB inlet barb 3/8-1/2 in. with 1/2 SB inlet and outlet barb
Pack for $178.66
 
09-747-16
View Documents
Cytiva
67177510
0.2/0.1 μm 3/8-1/2 in. 1/2 SB inlet and outlet barb 3/8-1/2 in. 1/2 SB inlet and outlet barb
Pack for $178.66
 
09-747-12
View Documents
Cytiva
67157502
0.2/0.2 μm 1/4-3/8 in. SB inlet barb 1/4-3/8 in. outlet plus filling bell barb
Pack for $182.94
 
09-747-5
View Documents
Cytiva
67147501
0.2/0.1 μm 1/4-3/8 in. SB inlet barb 1/4-3/8 in. SB outlet barb
Pack for $178.66
 
Description

Description

  • Two-layer filter for long life, high efficiency
  • Sterile, nonpyrogenic
  • Sterilizing grade filtration capsules
  • 0.2μm capsules meet bacterial retention per ASTM F838-83
  • Low-protein-binding membrane
  • Hydrophilic; low extractables
  • With inlet vent
  • Filter Media: Two layers polyethersulfone (PES)

Sterile filtration of biological and tissue culture solutions. Principally for aqueous solutions.

Specifications

Specifications

Polyethersulfone
930 cm2
Sterile filtration of biological and tissue culture solutions. Principally for aqueous solutions.
Sterile
SDS
Product Certifications