Please login to your online account to display your discounted pricing

Whatman™ pH Indicator Papers, CS Strips


Manufacturer: whatman™  WHT2627-990

 View more versions of this product

Catalog No. 09-876-19

  • / Pack of 200 Each
 

Description & Specifications

Specifications

Includes Integral color chart
pH Range 3.8 to 5.5
For Use With (Application) For the rapid determination of pH values has led to many applications in laboratories and industry
Graduations 0.2 or 0.3 pH
Quantity 200 Pack
Product Type pH Indicator and CS Test Strip