Please login to your online account to display your discounted pricing
Whatman™ pH Indicator Papers, CS Strips
$64.75 - $64.75
Specifications
For Use With (Application) For the rapid determination of pH values has led to many applications in laboratories and industry
View More Specs
Products
Catalog Number Mfr. No. Quantity Product Type pH Range Price Quantity    
09-876-19
whatman™
WHT2627-990
200 Pack pH Indicator and CS Test Strip 3.8 to 5.5 Pack of 200 Each for $64.75
09-876-21
whatman™
WHT2631990
200 Pack pH Indicator and CS Test Strip 9.5 to 12 Pack of 200 Each for $64.75
Description & Specifications
Specifications
For Use With (Application) For the rapid determination of pH values has led to many applications in laboratories and industry
  • Integral color chart, eight or more color segments graduated in 0.2 or 0.3pH units
  • Central segment changes color in solution; match color chart for value
  • Packs of 200; each strip is 0.43 × 30.93 in. (11 × 100mm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.