Whatman™ Optitran™ Nitrocellulose Membranes
Combine flexibility, strength, and excellent signal-to-noise ratios—ideal for experiments that involve stripping and reprobing
$335.94 - $687.75
Specifications
Binding Capacity 75 to 90μg/cm2
Material Nitrocellulose
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Dimensions (L x W) Format Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Format Pore Size Quantity Price Quantity  
36-100-5822 Whatman™
WHT10439116C
82mm (diameter) Circle 0.45μm 50/Pk. Pack of 50 Each for $335.94
36-100-5823 Whatman™
WHT10439126C
132mm (diameter) Circle 0.45μm 50/Pk. Pack of 50 Each for $687.75
Specifications
Description
  • 100% nitrocellulose supported by an inert polyester nonwoven membrane
  • Support does not affect transfer conditions or results
  • High sensitivity with very low nonspecific binding
  • Stringent washing and blocking conditions are not necessary
  • Binding capacity: 75 to 90μg/cm2
  • Autoclavable using liquid cool cycle
  • Also available in circles
Description & Specifications
Specifications
75 to 90μg/cm2
Nitrocellulose