Please login to your online account to display your discounted pricing

Whatman™ Nytran™ SuperCharge Membranes


Cast uniformly on both sides of a support matrix, providing excellent symmetry and allowing membranes to lie flat without curling

Manufacturer: whatman™  WHT10416216*

 View more versions of this product

Catalog No. 09-301-162

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Material Nylon
Format Circle
Binding Capacity >600μg/cm2
Pore Size 0.45μm
For Use With (Application) Suitable for isotopic and nonisotopic detection