Whatman™ 90mm Nuclepore™ Polycarbonate Track-Etched Membranes
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Manufacturer:  Whatman™ WHT111711
 View more versions of this product
Catalog No. 09-800-927
Specifications
Description
Membrane, Filter; Whatman; Nuclepore; Track-Etched polycarbonate (hydrophilic); Circle; High chemical, thermal resistance; Low protein binding, low extractables; Low, consistent ash, low tare weights; Pore Size: 2um; Dia.: 90mm
Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Description & Specifications
Specifications
2μm
90mm
Hydrophilic
Nuclepore Polycarbonate
Epifluorescence microscopy, environmental analysis, cell biology
High