Whatman™ 19mm Nuclepore™ Polycarbonate Track-Etched Membranes
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ 19mm Nuclepore™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Manufacturer:  Whatman™ WHT800281
 View more versions of this product
Catalog No. 05-715-115
Specifications
Description
Filters, Membrane; Whatman; Nuclepore track-etched polycarbonate; Circle; Dia.: 19mm; Pore size: 0.2μ; High flow rates; Low protein binding and low extractables; Excellent chemical and thermal resistance; 100/Pk.
Effective chemotaxis requires a delicate balance between cell adhesion and migration. Nucleopore Membranes combine PVP-free polypropylene with sharp, uniform cylindrical pores to achieve both.
  • Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Description & Specifications
Specifications
0.2μm
19mm
High
Polycarbonate film
Hydrophilic