Please login to your online account to display your discounted pricing
Whatman™ Nuclepore™ Polycarbonate Hydrophilic Membranes
Manufactured from high-quality polycarbonate film and track-etched for high pore size precision. Whatman™ Polycarbonate Hydrophilic Membranes combine high flow rates and chemical and thermal resistance with sharply defined pores, making them suitable for microscopy, cytology, parasitology, and certain air and oceanography analyses.
Manufacturer: whatman™  WHT111114
 View more versions of this product
Catalog No. 09-300-75
  • / Pack of 100 Each
Description & Specifications
Specifications
Wettability Hydrophilic
Material Nuclepore Polycarbonate
Membrane Type Track-etched
Quantity (No. per Case) 100Pack
Pore Size 8μm
Diameter (Metric) Outer 47mm
Offers a highly defined pore size cutoff in the sub-micron range, enabling particle deformity measurement and capture of extremely small particles.
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to render it hydrophilic
  • Low extractables, minimizing sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat surface for high particle visibility
Accessories
Accessories for Catalog Number: 0930075
Item Mfr. No. Item Description Price Per Unit Quantity