Whatman™ Mixed Cellulose Ester Membrane Filters
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
Manufacturer:  Whatman™ 10407370
 View more versions of this product
Catalog No. 09927778
Specifications
Description
Filter, Membrane; Whatman; Grade: ME25/41; Circles; Green; Mixed cellulose ester; For Membrane-Butler; Pore Size: 0.45um; 4.7cm; Grid: 3.1mm black
  • Sterile options available for critical applications
  • Contrast for easier particle detection
  • Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
  • Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
Description & Specifications
Specifications
Yes
Membrane butler
Hydrophilic
ME25
Medium
Yes,Yes
266°F
4.7cm
47mm
400
Green
Black
Mixed Cellulose Ester,Mixed Cellulose Ester (MCE)
0.45μm,0.45μm
0.31cm
Grid
130°C
Membrane Filter
Membrane Filter