Please login to your online account to display your discounted pricing

Whatman™ Microplate Seals

Self-sticking with inert adhesive. Whatman™ Microplate Seals prevent spillage and reduce sample evaporation.

$126.78 - $126.78

Specifications

Applications Used to control humidity and reduce evaporation of samples
Color Clear
Product Type Microplate Seal
View More Specs
Products
Catalog Number Mfr. No. Thickness Adhesive Material Price Quantity    

09-003-39

 
whatman™
77040001
0.05mm Cold, inert, self-sticking Polyester Pack of 100 Each for $126.78
Description & Specifications

Specifications

Applications Used to control humidity and reduce evaporation of samples
Color Clear
Product Type Microplate Seal

  • Controls humidity and reduces evaporation of samples.
  • Prevents spills and contamination.
  • Cold seals are self-sticking with inert adhesive.
  • Heat seals are available in a clear polyester or aluminum foil.
  • Heat seals are for polypropylene microplates only and are applied with heat and pressure.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.