Please login to your online account to display your discounted pricing
Whatman™ Microbiological Monitor
For use in the Membrane Filtration Technique to recover microorganisms from aqueous samples
$318.84 - $325.95
Products
Catalog Number Mfr. No. Price Quantity    
36-100-5927
whatman™
10497600
Pack of 50 Each for $318.84
09-805-344
maine manufacturing
10497500
Pack of 50 Each for $325.95
Description & Specifications
  • Applications include raw material to finished product sampling for beverages, foods, water and pharmaceuticals.