Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Whatman™ Glass Microfiber Extraction Thimbles, Grade 603G, without binder
Use for analysis of hot and acidic gases, including emission control and smoke tack gas monitoring. Whatman™ Glass Extraction Thimbles are designed to fit most Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.
Manufacturer: Cytiva (Formerly GE Healthcare Life Sciences) 2811308
Description
Use for analysis of hot and acidic gases, including emission control and smoke tack gas monitoring. Whatman™ Glass Extraction Thimbles are designed to fit most Soxhlet extraction units and can be used with solvents that are incompatible with cellulose thimbles.
- 603G borosilicate glass fiber thimble without binder
- Resistant to temperatures up to 500°C (932°F)
- Manufactured with walls as thin as 1–1.5 mm
- No binders are added
- Applications include: smoke stack gas monitoring, Soxhlet extraction, analyzing pesticide residues, determining oil/fat content of foods, analysis of oil and grease in solid wastes
Specifications
GE’s Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems. | |
Smoke stack gas monitoring. Soxhlet extraction. Analyzing pesticide residues. Determining oil/fat content of foods. Analysis of oil and grease in solid wastes. | |
603 g | |
500°C | |
1.18 x 3.15 in. | |
Borosilicate glass fibers |
30 mm | |
Soxhlet extractor | |
80 mm | |
Extraction Thimble | |
30 x 80mm |
Promotions
-
Fisherbrand
Get a Welcome Basket with $25 Cytiva Purchase
Spend $25 or more on Cytiva products and get a welcome gift basket at no additional cost.: 06/30/21