missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles

Designed to fit most Soxhlet extraction units. Whatman™ High Purity Glass Microfiber Soxhlet Extraction Thimbles are made of 100% borosilicate glass microfiber. They are well-suited to pollution monitoring when analysis of hot and acidic gases is required.

Manufacturer:  Whatman™ 2814199

 View more versions of this product

Catalog No. 09-657-1A


 

Specifications

Description

Thimble, Extraction; Whatman; Made of high-purity glass microfiber; HP grade; Binders/additives free; Suitable for both solvent extraction and air sampling; Withstands temp. up to 500 deg.C; Length: 90mm; I.D.: 19mm

  • Resistant to temperatures up to 500°C
  • Binder-free, no chemical additives
  • Designed to fit most commercially available Soxhlet extractors.
  • Effective when used with solvents incompatible with cellulose materials
  • Applications: analysis of hot and acidic gases during emission control and smoke stack gas monitoring, analysis of pesticide residues using Soxhlet extraction, determination of oil/fat content of foods, analysis of oil and grease in solid wastes>
Description & Specifications

Specifications

19mm
Soxhlet extraction units
90mm
500°C
1.7mm
Smoke stack gas monitoring, soxhlet extraction, analyzing pesticide residues, determining oil/fat content of foods, analysis of oil and grease in solid wastes
HP-GF
932°F
Extraction Thimble
Borosilicate glass