Cytiva Whatman™ HEPA-CAP 150
inlet 3/8 in. FNPT, outlet 3/8 in. FNPT
Supplier: Cytiva 67029500
Specifications
Autoclavable | |
Laminated Hydrophobically Treated Glass Microfiber | |
3/8 in. FNPT | |
1 Piece | |
5 Pieces |
HEPA-CAP 150, inlet 3/8 in. FNPT, outlet 3/8 in. FNPT (1 pc) | |
Polypropylene (PP) | |
4.1 bar (60 psi) | |
3/8 in. FNPT | |
150 mm |