Please login to your online account to display your discounted pricing

Whatman™ Grade Shark Skin Filter Paper for Technical Use

$105.07 - $1,018.11

Specifications

Format Sheet
Material Cellulose
Grade Shark skin
Thickness 170μm
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity    

09-800-962

 
whatman™
10537138
750 ft. 228.6m 21 in. 53cm Pack for $1,018.11

09-800-965

 
whatman™
10538877
10 in. 25.4cm 8 in. 20.32cm Pack for $105.07
Description & Specifications

Specifications

Format Sheet
Material Cellulose
Grade Shark skin
Thickness 170μm