Whatman™ Quartz Air Sampling Filters, Grade QM-A, Circle
Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols. Whatman™ Quartz Air Sampling Filters are sequentially numbered according to U.S. Environmental Protection Agency (EPA) standards and are suitable for most applications.
$240.63 - $1,608.77
Specifications
Type Circles
Airflow Rate 6.4 s/100 ml/in2
Grade QM-A
Material High-purity quartz
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Filter Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Filter Price Quantity  
09-916-6 Whatman™
1851101
Pack of 100 Each for $735.17
09-874-605 Whatman™
1851025
Pack of 100 Each for $240.63
09-874-607 Whatman™
1851090
Pack of 100 Each for $657.21
09-927-435 Whatman™
1851082
Pack of 100 Each for $583.02
09-874-608 Whatman™
1851150
Pack of 100 Each for $1,608.77
09-874-87 Accessories Whatman™
1851032
32mm Pack of 100 Each for $241.98
09-874-89 Accessories Whatman™
1851047
47mm Pack of 100 Each for $316.92
09-874-88 Whatman™
1851037
37mm Pack of 100 Each for $289.56
Specifications
Description
  • Temperature resistant up to 500°C
  • Binder-free
  • High loading capacity
  • Fast flow rates (air flow rate 6.4 s/100 ml/in2)
  • Used in many air sampling filter applications, including sampling of acidic gases, stacks, flues, and aerosols
  • Filters are suitable for PM-10 sampling and analysis
Description & Specifications
Specifications
Circles
6.4 s/100 ml/in2
QM-A
High-purity quartz