Please login to your online account to display your discounted pricing
Whatman™ Grade 8 Ruled Special-Purpose Filter Paper
Hardened grade for routine analysis of extraneous matter in food, beverage and pharmaceutical laboratories
Manufacturer: whatman™  WHT10347009*
 View more versions of this product
Catalog No. 09-924-385
  • / Pack of 100 Each
Description & Specifications
Specifications
Includes 5mm green rules are resistant to aqueous, non-aqueous solution
Product Type Ruled Special Purpose Filter Paper
Diameter (English) Inside 3.54 in.
Diameter (Metric) Inside 9cm
For Use With (Application) Qualitative analytical techniques to determine and identify materials
Material Cotton Linter