missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles

Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 595½ Pleated Circles provide extra surface area to increase total loading capacity.

$95.83 - $331.64

Specifications

Format Pleated circle
Thickness (Metric) 150μm
Particle Retention Medium to fine
Material Cellulose
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
09-924-165 Whatman™
WHT10311645
150mm Pack of 100 Each for $123.44
09-924-163 Whatman™
WHT10311643
110mm Pack of 100 Each for $95.83
09-924-164 Whatman™
WHT10311644
125mm Pack of 100 Each for $108.98
09-924-166 Whatman™
WHT10311647
185mm Pack of 100 Each for $150.10
09-924-167 Whatman™
WHT10311651
240mm Pack of 100 Each for $232.40
09-924-169 Whatman™
WHT10311653
320mm Pack of 100 Each for $331.64
09-924-168 Whatman™
WHT10311652
270mm Pack of 100 Each for $262.09
Specifications

Description

Folded filter is used for many routine analytical applications in various industries. Pleated paper enhances and eases filtering process. 
  • Saves time in repetitive or multiple analyses
  • Decreases overall filtration time; normal slow down due to particulate loading is eliminated
  • Increases total loading capacity
  • Maintains flow rate; contact with funnel is reduced, shape of filter is self-supporting
  • Pleating does not significantly affect technical data; same figures may be used with flat circles
  • Grade 595½ is thin, medium fast paper with medium to fine particle retention
  • Applications include:
    • Particle separation from food extracts
    • Filtration of solids from digested environmental sample
Description & Specifications

Specifications

Pleated circle
150μm
Medium to fine
Cellulose