Whatman™ Grade 520a Qualitative Folded Paper Filter Circle
For the filtration of viscous liquids and emulsions such as sweetened juices, spirits,syrups, resin solutions, oils or plant extracts
$191.80 - $499.85
Specifications
Format Circle
Flow Rate High
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity  
09-924-344 Whatman™
WHT10331456
500mm Pack of 100 Each for $499.85
09-924-343 Whatman™
WHT10331451
240mm Pack of 100 Each for $191.80
Specifications
Description
  • Great wet strength
  • Very high flow rate
  • Super-refined cellulose
Description & Specifications
Specifications
Circle
High