Please login to your online account to display your discounted pricing

Whatman™ Grade 114 Wet-Strengthened Qualitative Filter Paper: Circle

Coarse paper with creped or smooth surface

$27.83 - $452.31

Specifications

Format Circle
Grade 114
For Use With (Application) Use for coarse particles and gelatinous precipitates.
Particle Retention 25μm
Quantity 100 Pieces
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    

09-833D

  Accessories
whatman™
1114090
9cm Pack of 100 Each for $27.83

09-833F

  Accessories
whatman™
1114125
12.5cm Pack of 100 Each for $50.38

09-833G

  Accessories
whatman™
1114150
15cm Pack of 100 Each for $62.98

09-833H

  Accessories
whatman™
1114185
18.5cm Pack of 100 Each for $92.21

09-833J

  Accessories
whatman™
1114240
24cm Pack of 100 Each for $149.65

09-833M

  Accessories
whatman™
1114400
40cm Pack of 100 Each for $452.31
Description & Specifications

Specifications

Format Circle
Grade 114
For Use With (Application) Use for coarse particles and gelatinous precipitates.
Particle Retention 25μm
Quantity 100 Pieces
Thickness 190μm

  • Have high wet strength due to the addition of a small quantity of chemically stable resin

Cat. No. 09-833F is made to order, and therefore not returnable