missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Grade 0905 Qualitative Creped Filter Paper Sheets

Designed for applications ranging from the filtration of beverages to the purification of electroplating baths

$986.23 - $1,063.65

Specifications

Format Sheet
Grade 0905
Material Super-refined Cellulose
Flow Rate High
Pore Size (Metric) 12 to 25μm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
09-924-362 Whatman™
WHT10334987
580mm 580mm Pack of 500 Each for $1,063.65
09-924-361 Whatman™
WHT10334985
450mm 450mm Pack of 500 Each for $986.23
Specifications

Description

  • Very high filtration speed
  • Creped for coarse particles

Description & Specifications

Specifications

Sheet
Super-refined Cellulose
12 to 25μm
0905
High