Whatman™ Glass Mini-UniPrep™ G2 Syringeless Filter, 0.45um Pore Size and Normal Cap
Get advanced HPLC/UHPLC filter efficiency with the purity of glass
$490.54 - $4,674.93
Specifications
Pore Size 0.45μm
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Material Quantity Type Price Quantity  
Catalog Number Mfr. No. Material Quantity Type Price Quantity  
09-924-457 Whatman™
GN203NPUORG
PTFE 100/Pk. Syringeless Filter Each for $490.54
09-924-493 Whatman™
GN203NPUGMF
GMF 100/Pk. Syringeless Filter Pack of 100 Each for $531.61
09-924-494 Whatman™
GN203NPURC
RC 100/Pk. Syringeless Filter Pack of 100 Each for $531.61
09-924-500 Whatman™
GN203NPUGMFSP
GMF 100/Pk. Syringeless Filter w/ hand compressor Pack of 100 Each for $508.33
09-924-527 Whatman™
GN503NPUORG
PTFE 1000/Pk. Syringeless Filter Pack of 1000 Each for $4,674.93
09-924-458 Whatman™
GN203NPUORGSP
PTFE 100/Pk. Syringeless Filter w/ hand compressor Each for $528.75
09-924-461 Whatman™
GN203NPUAQU
PVDF 100/Pk. Syringeless Filter Each for $512.84
Specifications
Description
These all-in-one filters replace syringe filters, and speed up and simplify UHPLC/HPLC sample preparation. They provide a fast and easy way to remove particulates from samples being prepared for Ultra High Performance Liquid Chromatography (UHPLC)/High Performance Liquid Chromatography (HPLC) analysis. Due to the design of this next generation product, the filtered sample is stored within an integral borosilicate glass vial, minimizing the risk of leachables contaminating the sample. Standardizing UHPLC/HPLC sample preparation with Mini-UniPrep G2 ensures that every sample is filtered prior to analysis, and hence minimizes instrument downtime due to unfiltered samples. The time and money saved from cutting multiple consumables out of the sample preparation process provides immediate benefits for your laboratory.
  • Consists of an integral borosilicate glass autosampler vial, plunger with attached filter membrane, and normal cap
  • Allows you prepare samples faster compared to the syringe filter method
  • Glass construction of Mini-UniPrep G2 minimizes the risk of leachables contaminating the sample
  • Designed to be loaded directly into the autosampler
  • Visual indication that the sample has been filtered; minimizes instrument downtime due to unfiltered samples
  • With 0.45μm pore sizes to meet specific sample filtration requirements
  • Normal cap and with translucent housing
  • Sold in packs of 100, with or without a hand compressor and packs of 1000 vials
Description & Specifications
Specifications
0.45μm