Please login to your online account to display your discounted pricing

Whatman™ Glass Microfiber Filter Paper Grade 9

Glass microfiber filter paper

$228.04 - $465.60

Specifications

Format Circle
Grade GF 9
Material Glass Microfiber
Flow Rate 27 s/100mL/1.56cm2
Thickness 350μm
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity    

09-924-274

 
whatman™
WHT10370202
2 in. 50mm Pack of 200 Each for $228.04

09-924-276

 
whatman™
WHT10370206
4.33 in. 110mm Pack of 100 Each for $465.60

09-924-275

 
whatman™
WHT10370205
3.54 in. 90mm Pack of 100 Each for $314.00
Description & Specifications

Specifications

Format Circle
Grade GF 9
Material Glass Microfiber
Flow Rate 27 s/100mL/1.56cm2
Thickness 350μm

  • Filters used for various applications from fine and coarse particle analysis to prefilter membranes for crop protection residues
  • Flow rate: 27 s/100mL/1.56cm2