Please login to your online account to display your discounted pricing

Whatman™ Glass Microfiber Filter Paper Grade 8


Glass microfiber filter paper

Manufacturer: whatman™  WHT10370105

 View more versions of this product

Catalog No. 09-924-270

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Format Circle
Quantity 100/Pk.
For Use With (Application) Filters used for various applications from fine and coarse particle analysis to prefilter membranes for crop protection residues.
Flow Rate 12 s/100mL/1.56cm2
Thickness 350μm
Diameter (Metric) 90mm
Grade GF 6
Material Glass Microfiber