Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
missing translation for 'pleaseLogIn'

Whatman™ Glass Microfiber Filter Paper Grade 8

Glass microfiber filter paper

$194.29 - $356.84

Specifications

Material Glass Microfiber
Flow Rate 12 s/100mL/1.56cm2
Thickness 350μm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Diameter (Metric) Format Grade Price Qty  
Catalog Number Mfr. No. Quantity Diameter (Metric) Format Grade Price Qty  
09-924-272 Whatman™
WHT10370119
200/Pk. 47mm Circle GF 6 Pack of 200 Each for $194.29
09-924-273 Whatman™
WHT10370172
100/Pk. 60 x 90mm Rectangle GF 8 Pack of 100 Each for $356.84
09-924-270 Whatman™
WHT10370105
100/Pk. 90mm Circle GF 6 Pack of 100 Each for $314.00
Specifications

Description

  • For filtration of coarse particles and determination of PCB, DDE, DDT, furan, dioxin4
  • Flow rate:12 s/100mL/1.56cm2
  • Thickness: 350μm

Description & Specifications

Specifications

Glass Microfiber
12 s/100mL/1.56cm2
350μm