Whatman™ FTA™ Nucleic Acid Collection, Storage and Purification Cards: FTA Gene
Collect, store, and transport DNA at room temperature
$944.82 - $3,192.65
Specifications
For Use With (Application) Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
09-923-346 Whatman™
WHTWB120208
75μL Pack of 100 Each for $944.82
09-302-194 Whatman™
WHTWB120314
75μL Case of 10 Each for $3,192.65
Specifications
Description
Patented FTA technology simplifies the isolation, storage, and processing of nucleic acids from biological sources including whole blood, buccal cells, and plant material. Sample is applied to the FTA card and allowed to dry; the cells are lysed on contact. The released DNA is captured, stabilized, and protected from nucleases, oxidation, and UV damage.
  • DNA from blood stored at room temperature on FTA cards exhibits no loss in PCR efficiency after 22 years and counting
  • FTA Gene Card is housed in a chipboard frame for compatibility with most automatic dispensing/pipetting systems when used with the FTA Gene Card Tray (09-923-349)
Description & Specifications
Specifications
Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification