Please login to your online account to display your discounted pricing

Whatman™ Non-indicating FTA Classic Cards

Lyses cells on contact, denatures proteins and protects DNA from degradation. Whatman™ Non-indicating FTA Classic Cards contain chemical denaturants and a free radical scavenger keeping DNA tightly bound while proteins and inhibitors are washed from the matrix.

Manufacturer: whatman™  WB120312

 View all options for this product

Catalog No. 09-302-192


Description & Specifications


For Use With (Application) Forensics, transgenic identification, transfusion medicine/HLA typing, plasmid screening, food and agriculture testing, drug discovery, genomics, STR analysis, animal identification, whole genome amplification
For Use With (Equipment) Direct STR amplification
Format Four sample areas
No. of Samples 4
Product Type Nonindicating
Quantity 250 Pack
Sample Size 125μL per sample area
Volume (Metric) Sample 500μL max. per card

Collect, stabilize, process, transport, and archive blood samples.

  • Improves process efficiency by enabling direct amplification for STR analysis.
  • Ideal for colored samples, like blood.
  • Immediate stabilization and protection of nucleic acids due to FTA technology.
  • Different card options to support manual or automated processing for fast and efficient sampling.
  • Long term archiving of samples at room temperature due to FTA card technology.