Please login to your online account to display your discounted pricing

Whatman™ FTA™ Elute Cards


Provides room temperature DNA storage combined with simplified purification of DNA for multiple amplifications from a single sample. Whatman™ FTA™ Elute Cards contain a chaotropic salt keeping proteins tightly bound while DNA is eluted from the matrix.

Manufacturer: whatman™  WHTWB120401

 View more versions of this product

Catalog No. 09-301-230

  • / Pack of 25 Each
 

Description & Specifications

Specifications

For Use With (Application) STR Analysis
Volume (Metric) Sample 40μL
Salt Form Chaotropic
No. of Samples 4
Test Time 15 to 30 min.
Product Type FTA Card
Quantity 25 Pack
Storage Requirements Room Temperature