missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Grade 589/2 Quantitative Filter Paper

Retains medium fine precipitates using material with virtually no ash content. Whatman™ Grade 589/2 Quantitative Filter Paper is used when medium fine precipitates are being collected and papers must be ashed to support analysis.

Manufacturer:  Whatman™ WHT10300103

 View more versions of this product

Catalog No. 09-924-107


 

Specifications

Description

Paper, Qualitative filter; Whatman; Grade 589/2; Folded; Ash content: 0.01%; White ribbon; Particle retention in liquid: 4 to 12μm; Filtration speed: 70Hz; For determination of sand content in foodstuffs and more; Thickness: 40.5mm

Designed for gravimetric analysis

  • White Ribbon Filter for medium fine precipitates
  • Ashless
  • Medium filtration speed
  • For sand content measurement in foodstuffs
  • For flour grade determination
  • For aqueous suspension analysis

Description & Specifications

Specifications

100Pack
High-quality cotton linters
180μm
4 to 12μm
White Ribbon Filter
589/2
Circle
40.5mm
Medium