missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ EPM 2000 Air Sampling Filters

Used in high-volume PM-10 sampling equipment. Whatman™ EPM 2000 Air Sampling Filters are manufactured from 100% pure borosilicate glass. They enable detailed chemical analysis of trace pollutants with minimal interference.

Manufacturer:  Whatman™ 1882866

 View more versions of this product

Catalog No. 09-874-67


 

Specifications

Description

Filter, air sampling; Whatman; Borosilicate glass; Grade EPM 2000; Binder free; Airflow Rate: 4.7 s/100 ml/1.56 cm2; Basis weight: 85; Sheets; LxW: 203.2 x 254mm (8 x 10 in.)

  • Air retention efficiency of 99.95 percent of 0.3 μm size particulates
  • Selected by the U.S. Environmental Protection Agency (EPA) for use in a nationwide network of HiVol air samplers
  • Manufactured according to ISO9001:2008 QMS standards
  • Available in both sheet and circle design
  • Sheets are individually numbered to facilitate identification
  • Binder-free
Description & Specifications

Specifications

4.7 s/100 ml/1.56 cm2
203.2 x 254 mm (8 x 10 in)
EPM 2000
20.3cm
10 in.
High volume PM-10 air sampling equipment
Sheets
85
Sheet
8 in.
0.45mm
25.4cm
Borosilicate glass
Accessories