Please login to your online account to display your discounted pricing
Whatman™ Elutrap™ Electroelution Membranes
Designed to isolate nucleic acids and proteins from agarose or polyacrylamide gel slices
$402.13 - $557.00
Specifications
For Use With (Application) Eluting nucleic acids (14 bp to 150Kb) and proteins (larger than 3 to 5kD) from gel slices
For Use With (Equipment) Elutrap™ electroelution system
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    
09-301-141
whatman™
WHT10404090*
BT1 membrane Pack of 100 Each for $557.00
09-301-142
whatman™
WHT10404092*
BT2 membrane Pack of 100 Each for $402.13
Description & Specifications
Specifications
For Use With (Application) Eluting nucleic acids (14 bp to 150Kb) and proteins (larger than 3 to 5kD) from gel slices
For Use With (Equipment) Elutrap™ electroelution system
  • For most horizontal gel electrophoresis chambers
  • Efficient current flow
  • Handles up to four samples simultaneously
  • Purifies nucleic acids 14bp to 150Kb; proteins larger than 3 to 5kD
  • No salt cushions or special buffers required
  • Adjustable trap allows optimization of final sample volume