Cytiva Whatman™ Cyclopore™ Polycarbonate Membrane Filters
Ensures visibility of particles under the microscope with smooth membranes and low affinity for stains. Whatman™ Cyclopore™ Polycarbonate Membrane Filters can also reproduce microfiltration characteristics of track-etched membrane filters.
$147.91 - $298.00
Specifications
Material Polycarbonate
Hydrophobicity Hydrophilic
No. per Pack 100Pack
View More Specs
Products 9
Catalog Number Mfr. No. Diameter (Metric) Pore Size Color Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Pore Size Color Price Quantity  
099303J
View Documents Promotion Details
Cytiva
70602513
25 mm 5 μm Translucent
Pack of 100 Each for $164.15
09-930-5-F
View Documents Promotion Details
Cytiva
70604710
47 mm 1.0 μm Translucent
Pack of 100 Each for $255.00
09-930-5J
View Documents Promotion Details
Cytiva
70604713
47 mm 5 μm Translucent
Pack of 100 Each for $255.00
09-924-573
View Documents Promotion Details
Cytiva
70604714
47 mm 8 μm Translucent
Pack of 100 Each for $284.00
09924574
View Documents Promotion Details
Cytiva
70622513
25 mm 5 μm Clear
Pack of 100 Each for $182.10
09-927-987
View Documents Promotion Details
Cytiva
70914710
47 mm 1 μm Clear
Pack of 100 Each for $298.00
09927983
View Documents Promotion Details
Cytiva
70602514
25 mm 8 μm Translucent
Pack of 100 Each for $147.91
09927982
View Documents Promotion Details
Cytiva
70602511
25 mm 2 μm Translucent
Pack of 100 Each for $187.45
09-924-572
View Documents Promotion Details
Cytiva
70604712
47 mm 3.0 μm Translucent
Pack of 100 Each for $255.00
Description
Description
Ensures visibility of particles under the microscope with smooth membranes and low affinity for stains. Whatman™ Cyclopore™ Polycarbonate Membrane Filters can also reproduce microfiltration characteristics of track-etched membrane filters.
For filtration and particle detection. True surface capture provides easy examination of samples and short analysis times.
  • No particle shedding provides ultra clean filtrate
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • Low affinity for stains provides higher optical contrast and makes visibility under a microscope easy
  • Hydrophilic with low levels of nonspecific protein binding
  • Low tare weight
  • Autoclavable: 30 min. at 121°C
Specifications
Specifications
Polycarbonate
Hydrophilic
100Pack
SDS
missing translation for 'documents'
missing translation for 'documents'