Cytiva Whatman™ Cellulose Nitrate Membrane Filters
Manufactured under strictly controlled conditions. Whatman™ Cellulose Nitrate Membrane Filters have a noticeably improved flexibility and are made to tolerate abuse during handling, loading, and autoclaving without sacrificing integrity.
$256.00 - $391.61
Specifications
Format Disc
Material Cellulose Nitrate
Quantity 100 Pk.
Thickness (Metric) 105 to 140 μm
View More Specs
Products 13
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Pore Size Price Quantity  
09927283
View Documents
Cytiva
10400014
50 mm 12 μm
Pack of 100 Each for $363.00
09927284
View Documents
Cytiva
10400106
25 mm 8 μm
Pack of 100 Each for $256.00
09-927-285
View Documents
Cytiva
10400112
47 mm 8 μm
Pack of 100 Each for $391.61
09927287
View Documents
Cytiva
10400206
25 mm 5 μm
Pack of 100 Each for $256.00
09-927-289
View Documents
Cytiva
10400212
47 mm 5 μm
Pack of 100 Each for $391.61
09927293
View Documents
Cytiva
10400214
50 mm 5 μm
Pack of 100 Each for $363.00
09-927-370
View Documents
Cytiva
10401312
47 mm 0.2 μm
Pack of 100 Each for $318.00
09927371
View Documents
Cytiva
10401314
50 mm 0.2 μm
Pack of 100 Each for $331.00
09927373
View Documents
Cytiva
10402014
50 mm 0.1 μm
Pack of 100 Each for $335.00
09-927-372
View Documents
Cytiva
10402012
47 mm 0.1 μm
Pack of 100 Each for $328.00
09927920
View Documents
Cytiva
7191005
50 mm 1.2 μm
Pack of 100 Each for $350.00
09927282
View Documents
Cytiva
10400012
47 mm 12 μm
Pack of 100 Each for $343.00
09927286
View Documents
Cytiva
10400114
50 mm 8 μm
Pack of 100 Each for $363.00
Description
Description
Among the strongest of their type available, as measured and compared by burst pressure tests.
  • Narrow pore size distribution for improved surface capture and analysis.
  • Low levels of extractables to ensure sample integrity.
  • High strength and flexibility
  • Increased temperature stability
  • Reduced shrinkage
Specifications
Specifications
Disc
Cellulose Nitrate
100 Pk.
105 to 140 μm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'