missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Binder-Free Glass Microfiber Filters GF/F Circles

Recommended by the EPA when the Toxicity Characteristic Leaching Procedure 1311 was developed. Whatman™ Binder-Free Glass Microfiber Filters GF/F Circles remain a popular and prominent component for measuring landfill leachate composition with a TCLP test.

Manufacturer:  Whatman™ 1825090

 View more versions of this product

Catalog No. 09-874-73


 

Specifications

Description

Filter, Glass microfiber; Whatman; Type GF/F; Binder-free; Retain fine particles down to 0.7μm; Very rapid flowrate; Very high loading capacity; For filtering finely precipitated proteins; Diameter: 9.0cm

  • High loading capacity
  • 0.6–0.8 µm nominal particle retention
  • Medium filtration speed
Applications:
  • DNA binding and purification
  • Filtering finely precipitated proteins
  • Filtering biochemical solutions, fluids, and nucleic acids
Description & Specifications

Specifications

Yes
19 sec./100mL/in.2
GF/F
932°F
0.016 in.
9cm
Borosilicate Glass
3.5 in.
Circle
0.7μm
500°C
420μm
Effective in filtering finely precipitated proteins, can be used with type GF/D as a prefilter for clarification of extremely difficult biochemical solutions and nucleic acids
Accessories
Promotions

Promotions