Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A Sheets
Combines high loading capacity with fine particle retention. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A Sheets are ideal for gravimetric determination of airborne particulates, stack sampling and absorption methods of air pollution monitoring.
Manufacturer:  Whatman™ 1820866
 View more versions of this product
Catalog No. 09-927-858
Specifications
Description
Filter; Binder-free glass microfiber; GE Whatman; Type GF/A; Sheet; Fast, high loading; For water and air pollution monitoring; Particle retention: 1.6μm; 8 x 10 in. (20.3 x 25.4cm)
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500ºC. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • High flow rate
  • Good loading capacity
  • Water pollution effluent monitoring
  • Filtering water, algae, bacterial cultures
  • Foodstuff analyses
  • Protein filtration
  • Radioimmunoassay of weak β-emitters
Description & Specifications
Specifications
13 sec./100mL
GF/A
Sheet; L x W: 20 x 25.4cm
500°C
0.26mm
Glass Microfiber
Sheet
1.6μm
932°F
0.01 in.
Water pollution effluent monitoring, filtering water, algae, bacterial cultures, foodstuff analyses, protein filtration, radioimmunoassay of weak beta-emitters, gravimetric determination of airborne particulates