Please login to your online account to display your discounted pricing

Whatman™ Autovial™ 5mL Syringeless Filters


Replaces syringe-coupled devices with single filter units for one-time use. Whatman™ Autovial™ 5mL Syringeless Filters are pre-assembled filtration devices for removing particulates from samples.

Manufacturer: whatman™  WHTAV115NPUAQU

 View more versions of this product

Catalog No. 09-921-15

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Material Polyvinylidene Fluoride (PVDF)
Sterile No
Diameter (Metric) 13mm
Quantity 50/pk.
Volume (Metric) <=5mL
Pore Size 0.45μm
Type Syringeless Filter

Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

 

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Remove particulates from samples up to 5mL in volume
  • Filtration area: 1.7cm2

 

Glass Microfiber (GMF)

  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
  • Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
  • Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
  • Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples