missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Whatman™ Autovial™ Syringeless Filters: 12mL Capacity

Faster sample handling, up to 12mL in volume

Manufacturer:  Whatman™ AV125UNAO

 View more versions of this product

Catalog No. 09-917A


 

Specifications

Description

Filter, Syringeless; Whatman; Autovial; Graduated barrel; Built-in air purge; Diameter: 25mm; w/glass microfilter preFilter, Handles samples up to 12mL in volume; Membrane type: nylon; Pore size: 0.45um; 50/Pk.

Replace syringe-coupled devices with single filter units for one-time use. Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.

  • Preassembled units consist of a filter barrel and a plunger
  • Built-in air purge minimizes holdup
  • Integrated construction eliminates filter pop-off
  • Support stand protects slip-Luer tip, recessed to reduce chance of contamination
  • Handle samples up to 12mL in volume
  • All have an integral 25mm diameter filter in barrel (graduated in 1mL intervals) with polyethylene plunger
  • Filtration area: 3.0cm2
  • Glass microfiber prefilter ensures higher flow with less clogging
  • Units with NYL, PTFE, and PVDF membranes are available sterile

Cellulose Acetate (CA)

  • Low nonspecific protein binding, high loading capacity; for biologicals
Glass Microfiber (GMF)
  • High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
Nylon (NYL)
  • Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
Polypropylene (PP)
  • For aqueous and organic solutions; ideal for aggressive samples and high debris
Polytetrafluoroethylene (PTFE)
  • Hydrophobic; for samples >50% organic solvents
Polyvinylidene Fluoride (PVDF)
  • Low protein binding; for aqueous samples

Description & Specifications

Specifications

Syringeless Filter
Nylon (NYL)
50/pk.
No
25mm
<=12mL
0.45μm