missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ Autovial™ Syringeless Filters: 12mL Capacity, Nonsterile
Faster sample handling, up to 12mL in volume
$441.00 - $6520.00
Specifications
Sterility | Non-sterile |
---|---|
Type | Syringeless Filter |
Volume (Metric) | ≤12 mL |
Diameter (Metric) | 25 mm |
Catalog Number | Mfr. No. | Material | Pore Size | Sterility | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Pore Size | Sterility | Price | Quantity | |||
05-713-720
|
Cytiva
AV525UAQU |
PVDF | 0.45 μm | Non-sterile |
Pack of 1000 Each for $6,520.00
|
|
|||
09-916A
|
Cytiva
AV125UAQU |
PVDF | 0.45 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-916B
|
Cytiva
AV125EAQU |
PVDF | 0.2 μm | Non-sterile |
Pack of 50 Each for $443.00
|
|
|||
09-917A
|
Cytiva
AV125UNAO |
Nylon | 0.45 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-917B
|
Cytiva
AV125ENAO |
Nylon | 0.2 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-918B
|
Cytiva
AV125EORG |
PTFE | 0.2 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-918A
|
Cytiva
AV125UORG |
PTFE | 0.45 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-919
|
Cytiva
AV125UGMF |
Glass Microfiber | 0.45 μm | Non-sterile |
Pack of 50 Each for $441.00
|
|
|||
09-921-4C
|
Cytiva
AV125UCA |
Cellulose Acetate | 0.45 μm | Non-sterile |
Pack of 50 Each for $458.00
|
|
|||
22-042627
|
Cytiva
AV525UORG |
PTFE | 0.45 μm | Non-sterile |
Pack of 1000 Each for $6,520.00
|
|
|||
09-800-989
|
Cytiva
AV525UNAO |
Nylon | 0.45 μm | Non-sterile |
Pack of 1000 Each for $6,520.00
|
|
|||
Description
Replace syringe-coupled devices with single filter units for one-time use. Often used for removing particulates from potentially hazardous samples. Pour or pipet sample into filter barrel, insert plunger and compress to filter into autosampler vial.
- Preassembled units consist of a filter barrel and a plunger
- Built-in air purge minimizes holdup
- Integrated construction eliminates filter pop-off
- Support stand protects slip-Luer tip, recessed to reduce chance of contamination
- Handle samples up to 12mL in volume
- All have an integral 25mm diameter filter in barrel (graduated in 1mL intervals) with polyethylene plunger
- Filtration area: 3.0cm2
- Glass microfiber prefilter ensures higher flow with less clogging
- Units with NYL, PTFE, and PVDF membranes are available sterile
Cellulose Acetate (CA)
- Low nonspecific protein binding, high loading capacity; for biologicals
- High loading capacity, binder-free, fast flowrates; for clarifying aqueous or organic solutions, pH 3 to 8
- Inherently hydrophilic; for clarifying aqueous and organic samples, pH 3 to 10
- For aqueous and organic solutions; ideal for aggressive samples and high debris
- Hydrophobic; for samples >50% organic solvents
- Low protein binding; for aqueous samples
Specifications
Non-sterile | |
Syringeless Filter | |
≤12 mL | |
25 mm |