Please login to your online account to display your discounted pricing
Whatman™ Aqueous IFD™ and Solvent IFD™
Manufacturer: whatman™  WHT67265002
 View more versions of this product
Catalog No. 09-703-90
  • / Pack of 10 Each
Description & Specifications
Specifications
Product Type Aqueous IFD
For Use With (Application) Whatman Aqueous and Solvent IFD Inline Filter Degassers
Pore Size 0.2μm
Housing Material Polypropylene