Please login to your online account to display your discounted pricing

Whatman™ Anotop™ 25 Sterile Syringe Filters


Remove virtually 100% of particulates larger than pore size

Manufacturer: whatman™  68092102

 View more versions of this product

Catalog No. 09-926-13

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Type Syringe Filter
Diameter (Metric) 25mm
Material Anopore Inorganic Membrane (alumina matrix)
Pressure Rating 75psi
Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Hold up Volume (After Air Purge) <150μL
Disposable Yes
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Temperature (English) 113°F
Temperature (Metric) 45°C
Quantity 50/Pk.
Pore Size 0.02μm
Sterile Yes
Tip Material Polypropylene
Pressure Rating (Metric) 5.2kg/cm