Please login to your online account to display your discounted pricing

Whatman™ Anotop™ 25 Sterile Syringe Filters

Remove virtually 100% of particulates larger than pore size

$1,229.87 - $1,328.70

Specifications

Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Diameter (Metric) 25mm
Disposable Yes
Hold up Volume (After Air Purge) <150μL
Inlet Style Female Luer Lok
View More Specs
Products
Catalog Number Mfr. No. Pore Size Price Quantity    

09-926-11

 
whatman™
68092122
0.2μm Pack of 50 Each for $1,328.70

09-926-13

 
whatman™
68092102
0.02μm Pack of 50 Each for $1,229.87

09-926-12

 
whatman™
68092112
0.1μm Pack of 50 Each for $1,328.70
Description & Specifications

Specifications

Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Diameter (Metric) 25mm
Disposable Yes
Hold up Volume (After Air Purge) <150μL
Inlet Style Female Luer Lok
Material Anopore Inorganic Membrane (alumina matrix)
Outlet Style Male Luer Slip
Pressure Rating 75psi
Pressure Rating (Metric) 5.2kg/cm
Quantity 50/Pk.
Sterile Yes
Temperature (English) 113°F
Temperature (Metric) 45°C
Tip Material Polypropylene
Type Syringe Filter

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow

Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders. Sterile units are individually packaged to avoid contamination.