Please login to your online account to display your discounted pricing

Whatman™ Anotop™ 25 Plus Non-Sterile Syringe Filters


Hold-up volume: <30μL (10mm size), <200μL (25mm)

Manufacturer: whatman™  WHT68094022

 View more versions of this product

Catalog No. 09-926-20

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Housing Material Polypropylene
Hold up Volume (After Air Purge) <200μL
Pore Size 0.2μm
Type Syringe Filter
Quantity 50/Pk.
Diameter (Metric) 25mm
Pressure Rating 100psi