GE Healthcare Whatman™ Anotop™ 25 Non-Sterile Syringe Filters
Remove virtually 100% of particulates larger than pore size
$970.82 - $3,548.94
Specifications
Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Diameter (Metric) 25mm
Type Syringe Filter
Disposable Yes
Housing Material Polypropylene
View More Specs
Products 4
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
Catalog Number Mfr. No. Pore Size Quantity Price Quantity  
09-926 View Documents GE Healthcare
68092024
0.2μm 200/Pk. Pack of 200 Each for $3,548.94
09-926-8 View Documents GE Healthcare
68092012
0.1μm 50/Pk. Pack of 50 Each for $1,066.28
09-926-7 View Documents GE Healthcare
68092022
0.2μm 50/Pk. Pack of 50 Each for $970.82
09-926-9 View Documents GE Healthcare
68092002
0.02μm 50/Pk. Pack of 50 Each for $1,182.32
Description
Description
Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).
  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow
Order Info
Hexagonal, thermal-welded polypropylene housing is free of adhesives and binders.
Specifications
Specifications
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water
Syringe Filter
Polypropylene
Anopore Inorganic Membrane (alumina matrix)
75psi
45°C
25mm
Yes
Female Luer Lok
Male Luer Slip
Non-sterile