Please login to your online account to display your discounted pricing

Whatman™ Anotop™ 25 Non-Sterile Syringe Filters


Remove virtually 100% of particulates larger than pore size

Manufacturer: whatman™  WHT68092002

 View more versions of this product

Catalog No. 09-926-9

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Disposable Yes
Housing Material Polypropylene
Hold up Volume (After Air Purge) <150μL
Pore Size 0.02μm
Filtration Area (Metric) 4.78cm sq.
Inlet Style Female Luer Lok
Outlet Style Male Luer Slip
Sterile No
Type Syringe Filter
Temperature (Metric) Max 45°C
Quantity 50/Pk.
Diameter (Metric) 25mm
Material Anopore Inorganic Membrane (alumina matrix)
Pressure Rating 75psi
Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water