Please login to your online account to display your discounted pricing

Whatman™ Anotop™ 10mm Syringe Filters


Removes virtually 100% of particulates larger than pore size. Whatman™ Anatop™ 10mm Syringe Filters include a highly defined capillary pore structure and narrow pore-size distribution, enabling a high level of consistency in particle retention.

Manufacturer: whatman™  WHT68091002

 View more versions of this product

Catalog No. 09-926-3

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Volume (Metric) Up to 10mL
Filtration Area (Metric) 0.78cm2
Inlet Style Female Luer Lock
Sterile Nonsterile
Quantity 50Pack
Pressure Rating 6.9bar (100psi)
Disposable Yes
Housing Material Polypropylene
Hold up Volume (After Air Purge) <20μL
Pore Size 0.02μm
Outlet Style Male Luer Slip
Type Syringe Filter
Temperature (Metric) Max 45°C
Diameter (Metric) 10mm
Material Anopore Inorganic Membrane (alumina matrix)
Applications HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water

Compatible with aqueous suspensions and a wide range of organic solvents. Handle volumes up to 5mL (10mm diameter filter) or 100mL (25mm diameter filter).

  • Nondeformable, honeycombed Anopore™ membrane
  • Inherently hydrophilic membrane is free of adhesives and surfactants
  • Low protein binding, low fluid retention for maximum sample recovery
  • Unidirectional flow