Whatman™ Anodisc™ Filter Membranes
Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.
Manufacturer:  Whatman™ 68096012
 View more versions of this product
Catalog No. 09-926-33
Specifications
Description
Filters, Membrane; Whatman Anodisc; Pore Size: 0.1μm; Diameter: 25mm; Supported; With Annular Ring; Pack of 50
Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.
  • Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
  • No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
  • Extremely low protein binding minimizes sample loss.
  • Virtually transparent when wet making it highly suitable for microscopy studies.
Description & Specifications
Specifications
0.1μm
25mm
No
Hydrophilic
Aluminum oxide
50Pack
Polypropylene support ring
Anodisc 25
Accessories