Please login to your online account to display your discounted pricing
Whatman™ Anodisc™ Filter Membranes
Constructed with precision pore structure and narrow pore size distribution. Whatman™ Anodisc™ Filter Membranes ensure a high level of particle removal efficiency. Captures microorganisms and particulates on the surface of the membrane for subsequent analysis.
Manufacturer: whatman™  WHT68095012
 View more versions of this product
Catalog No. 09-926-36
  • / Pack of 50 Each
Description & Specifications
Specifications
Hydrophobicity Hydrophilic
Autoclavable No
Material Aluminum oxide
Includes Polypropylene support ring
Membrane Type Anodisc 47
Quantity (No. per Case) 50Pack
Pore Size 0.1μm
Diameter (Metric) Outer 47mm
Anopore inorganic membrane circles available with or without support ring contain no lateral crossovers between individual pores.
  • Wide solvent compatibility reduces the need to stock a variety of membranes in the laboratory.
  • No additives used in the manufacturing process ensures minimal extractables and no sample contamination.
  • Extremely low protein binding minimizes sample loss.
  • Virtually transparent when wet making it highly suitable for microscopy studies.
Accessories
Accessories for Catalog Number: 0992636
Item Mfr. No. Item Description Price Per Unit Quantity